Description
- Available in different sizes
- Made out of stainless steel
Specifications
Specifications
| Material | Stainless Steel |
| Autoclavable | Autoclavable |
| For Use With (Application) | Buerkle Vampire Sampler |
| For Use With (Equipment) | 500 mL Bottle |
| Diameter (Metric) | 84 mm |