Learn More
Cleaver Scientific Basic Gel Documentation Hood
Product Code. 16684193 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Model:
GDH-BASIC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
16684193 GDH-BASIC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16684193
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 08-12-2026
A low cost, simple to use gel documentation hood for use with a mobile phone. Reduces background from ambient light when using blue light transilluminators.
  • Place the hood over a Blue light Transilluminator
  • Use a phone to zoom in and take an image
  • Transfer the images from phone to analysis software
  • Ideal for education use or where low costs are important
Specifications
For Use With (Equipment) Mobile Phone
Model GDH-BASIC
Product Type Gel Documentation Hood