Learn More
JULABO™ Base Tray for Test Tube Racks
Product Code. 10051122
Change view
Click to view available options
For Use With (Equipment):
SW22, SW23 shaking water bath
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10051122 SW22, SW23 shaking water bath Each
1 options
Product Code. 10051122
Brand: JULABO™ 8970369
Only null left
Add to basket
Add to basket
For assembling max. 4 test tube racks. This accessory is suitable for JULABO™ SW22, SW23.
Specifications
For Use With (Equipment) SW22, SW23 shaking water bath
Type Basic Tray
Holds Assembling max. 4 test tube racks