Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Product Code. 10024771 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Calibration Module
Deionized Water Feed (Low Organics, Type I Water)
Deionized Water Feed (Ultralow Organics, Type I Water)
Low Water Level Pump Protector
R/O and Distilled Feed (Low Organics, Type I Water)
R/O and Distilled Feed (Ultralow Organics, Type I Water)
Unit Size:
Each
Product Code. Type unitSize
10024771 Deionized Water Feed (Ultralow Organics, Type I Water) Each
10034781 R/O and Distilled Feed (Ultralow Organics, Type I Water) Each
10186251 R/O and Distilled Feed (Low Organics, Type I Water) Each
10739692 Deionized Water Feed (Low Organics, Type I Water) Each
12740896 Low Water Level Pump Protector Each
10688333 Calibration Module Each
6 options
Product Code. 10024771
Brand: Thermo Scientific™ D50281
Only null left
Add to basket
Add to basket
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Nanopure Water Purifications System
Type Deionized Water Feed (Ultralow Organics, Type I Water)