Learn More
Thermo Scientific™ Barnstead™ Hose Nipple Cartridges
Product Code. p-4531456 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
1/2 Size Mixed-Bed
1/2 Size Mixed-Bed with Oxygen Removal
2/3 Size High Capacity
2/3 Size Mixed-Bed and Organic Removal
Full Size High Capacity
Full Size Macroreticular, Cation Removal
Full Size Organic Removal
Full Size Oxygen Removal
Full Size Pretreatment and Scale Eliminator
Full Size Ultrapure
Full Size Ultrapure with Organic Removal
Hose Nipple Cartridge Holder
Hose nipple cartridge, Full Size Cation Removal
Resin Type:
Activated Carbon
Cation Exchange
Macroreticular Cation Exchange
Mixed-bed Ion Exchange with Activated Carbon
Mixed-bed and Strong Cation Exchange, Activated Charcoal
Strong Anion Exchange
Two Bed Ion Exchange
Two Bed Ion Exchange with Strong Cation Exchange
Ultrapure Ion Exchange
Ultrapure Mixed Bed Ion Exchange
Ultrapure Mixed Bed with Strong Anion Exchange
Unit Size:
1 unit
Each
Product Code. Description Resin Type unitSize
15332168 1/2 Size Mixed-Bed Ultrapure Ion Exchange 1 unit
11746299 1/2 Size Mixed-Bed with Oxygen Removal Ultrapure Mixed Bed with Strong Anion Exchange Each
11810860 Full Size High Capacity Two Bed Ion Exchange Each
10423865 Full Size Ultrapure Ultrapure Mixed Bed Ion Exchange Each
12770746 Full Size Oxygen Removal Strong Anion Exchange Each
10771834 Full Size Organic Removal Activated Carbon Each
10311285 Hose nipple cartridge, Full Size Cation Removal Cation Exchange Each
15392178 Full Size Macroreticular, Cation Removal Macroreticular Cation Exchange 1 unit
12790746 Full Size Ultrapure Ultrapure Ion Exchange Each
11502954 Full Size Pretreatment and Scale Eliminator Mixed-bed and Strong Cation Exchange, Activated Charcoal Each
15302188 Full Size Ultrapure with Organic Removal Mixed-bed Ion Exchange with Activated Carbon 1 unit
11850850 2/3 Size High Capacity Two Bed Ion Exchange with Strong Cation Exchange Each
11512954 2/3 Size Mixed-Bed and Organic Removal Mixed-bed Ion Exchange with Activated Carbon Each
10034313 Hose Nipple Cartridge Holder - Each
14 options
Product Code. 15332168
Brand: Thermo Scientific™ D50220
Only null left
Add to basket
Add to basket
Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.
Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.
  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8″ barb hose connections on each end D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8″ ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges
TRUSTED_SUSTAINABILITY
Specifications
Description 1/2 Size Mixed-Bed
For Use With (Equipment) Barnstead Water Purification System
Particle Retention 430 g
Type Cartridge, Hose Nipple
Flow Rate Up to 75 L/hr.
For Use With (Application) Ultrapure Ion Exchange
Resin Type Ultrapure Ion Exchange