Learn More
Thermo Scientific™ Barnstead™ GenPure™ xCAD Water Purification System
Product Code. 13062953 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Discontinued; replacement product is 50136146
Discontinued; replacement product is 50136149
Discontinued; replacement product is 50136150
Discontinued; replacement product is 50136152
Discontinued; replacement product is 50136153
Discontinued; replacement product is 50136165
Discontinued; replacement product is 50136167
Discontinued; replacement product is 50136170
Discontinued; replacement product is 50136171
Discontinued; replacement product is 50136172
Unit Size:
Each
Product Code. Description unitSize
13062953 Discontinued; replacement product is 50136146 Each
13052953 Discontinued; replacement product is 50136150 Each
13585360 Discontinued; replacement product is 50136152 Each
13032953 Discontinued; replacement product is 50136149 Each
13042953 Discontinued; replacement product is 50136153 Each
13072953 Discontinued; replacement product is 50136165 Each
13092953 Discontinued; replacement product is 50136167 Each
13575270 Discontinued; replacement product is 50136170 Each
13082953 Discontinued; replacement product is 50136171 Each
13002963 Discontinued; replacement product is 50136172 Each
10 options
Product Code. 13062953
Brand: Thermo Scientific™ 50131298
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is no longer available. The recommended replacement is Thermo Scientific™ Barnstead™ GenPure™ xCAD Plus.
This item is no longer available and has been replaced by the next generation of Thermo Scientific™ Barnstead™ Water Purification Systems. The recommended replacement for this product is Barnstead™ GenPure™ xCAD Plus.
Gain flexibility and control with the compact remote dispenser in the Thermo Scientific™ Barnstead™ GenPure™ xCAD Water Purification System. This system delivers ultrapure water with consistent quality for the most demanding and sensitive applications. The remote dispenser can be wall- or bench-mounted. Ideal for molecular biology, microbiology and analytical chemistry applications, the system features integrated feed water monitoring, quick-connect cartridge replacement and exact dosing.
Integrated feed water monitoring:
  • Additional measuring cell monitors feed water conductivity.
  • If the quality of the feed water drops below the set value, a fault message is immediately displayed.
UV Photo-oxidation 185/254 nm:
  • High performance UV assembly reliably reduces the content of microorganisms and their metabolites.
Ready to use:
  • Feed water pressure switch, wall bracket, UV lamp, ultrapure cartridge and sterile filter are all included in one part number.
Measurement of conductivity/resistivity:
  • Conductivity cells are carefully calibrated prior to each measurement via built-in reference resistance with cell constants at 0.01 cm-1.
  • Temperature measurements are made by a platinum chip sensor with ±0.1°C accuracy.
Quick-connect cartridge replacement:
  • Aquastop quick-connect capability enables cartridge replacement within seconds even mid-operation.
Exact dosing:
  • Electronic dosing allows fully automatic volume control of 0.01-65L with <0.5% accuracy.
GLP-compliant documentation:
  • Real-time clock and code-protected operating system prevent unauthorized changes to system settings.
  • RS-232 interface with adjustable send interval allows safe data transfer of all measured data, faults, date and time to a PC computer or log printer.
  • Digital microprocessor control automatically monitors and stores faults from the last four weeks.
  • USP-compliant conductivity measurement with temperature compensation can be switched on or off.
TRUSTED_SUSTAINABILITY
Specifications
Conductivity 0.055 µS/cm
Description Discontinued; replacement product is 50136146
Bacterial Content <1 CFU/mL
Endotoxin Concentration 0.001 EU/mL
Feedwater Connector R 3/4 in.
Feed Water Monitoring Yes
Feedwater Source Pretreated
Flow Rate Up to 2 L/min.
Remote Dispenser Yes
Resistivity 18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring Yes
Total Organic Carbon 1 to 5 ppb
Type Water Purification System
Feedwater Pressure 1 to 6 bar
Operating Pressure 2 to 6 bar
Particles >0.2µM/mL <1 μM/mL
Power Consumption 0.1 kW
Temperature Range (Metric) 2°C to 35°C
Width (Metric) 372 mm
Height (Metric) 615 mm
Depth (Metric) 330 mm
Net Weight (Metric) 25 kg
Electrical Requirements 90/240 V 50/60 Hz
Voltage 90/240 V
Frequency 50/60 Hz