Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Description
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
Specifications
Specifications
| Description | Cartridge Kit |
| For Use With (Equipment) | Barnstead Easypure RoDi |
| Type | Cartidge Kit |