Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Specifications
Specifications
| Feedwater Source | Tap or Pretreated |
| Flow Rate | 3.8 |
| For Use With (Application) | Water purification cartridge |
| For Use With (Equipment) | Nanopure II, Older Nanopure Systems |
| Includes | One 09-034 high capacity ion exchange, 09-034-104 Organic/colloid and two 09-034-3 Ultrapure™ ion exchange cartridges |
| Model | D3804 |
| Pressure | 30 psi |
| Type | Cartridge Kit |
| Length (English) | 17 in. |
| Length (Metric) | 4.18 cm |
| Show More |