Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Product Code. 11756259 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Nanopure II, Older Nanopure Systems
Nanopure Infinity Systems
Nanopure Quad-style System
Feedwater Source:
Deionized, Tap
Pretreated
Reverse Osmosis, Distilled
Tap
Tap or Pretreated
Model:
D3804
D3805
D4801
D4802
D50254
D50255
D50256
D50257
D5026
missing translation for 'unitSize'
Each
Product Code. 11756259
missing translation for 'mfr': Thermo Scientific™ D4802
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
For Use With (Application) Water purification cartridge
For Use With (Equipment) Nanopure Quad-style System
Includes One 09-034-5C Macropure™ Pretreatment, 09-034 high capacity ion exchange, 09-050-276 Ultrapure™ and 09-050-267 Organicfree cartridges
Model D4802
Type Cartridge Kit