Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Product Code. 10256704 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Nanopure II, Older Nanopure Systems
Nanopure Infinity Systems
Nanopure Quad-style System
Feedwater Source:
Deionized, Tap
Pretreated
Reverse Osmosis, Distilled
Tap
Tap or Pretreated
Model:
D3804
D3805
D4801
D4802
D50254
D50255
D50256
D50257
D5026
Unit Size:
Each
Product Code. For Use With (Equipment) Feedwater Source Model unitSize
10256704 Nanopure Infinity Systems Deionized, Tap D50254 Each
11946092 Nanopure Quad-style System Pretreated D5026 Each
11860810 Nanopure Infinity Systems Reverse Osmosis, Distilled D50255 Each
11756259 Nanopure Quad-style System Tap D4802 Each
10774267 Nanopure II, Older Nanopure Systems Tap or Pretreated D3805 Each
15129583 Nanopure II, Older Nanopure Systems Tap or Pretreated D3804 Each
11840810 Nanopure Infinity Systems Reverse Osmosis, Distilled D50257 Each
11830810 Nanopure Infinity Systems Deionized, Tap D50256 Each
11746259 Nanopure Quad-style System Tap D4801 Each
9 options
Product Code. 10256704
Brand: Thermo Scientific™ D50254
Only null left
Add to basket
Add to basket
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Deionized, Tap
For Use With (Application) Water purification cartridge
For Use With (Equipment) Nanopure Infinity Systems
Includes One 09-050-226 Tap/DI Feed Pretreatment, 09-050-246 Organicfree cartridge, two 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters
Model D50254
Type Cartridge Kit