Learn More
Barium titanate (IV), Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Barium titanate (IV) |
| Melting Point | 1625°C |
| CAS | 12047-27-7 |
| Quantity | 50 g |
| Density | 6.08 g/cm³ at 20°C |
| Synonym | Barium titanate (IV), Barium metatitanate, Barium titanate (IV) |
| Molecular Weight (g/mol) | 233.19 |
| Grade | Not Available |