missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Hydroxide Octahydrate, Supelco™
Description
We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives. Barium hydroxide octahydrate supports greener technologies by serving as an efficient, potentially reusable base catalyst for biodiesel production from non‐edible oils
Specifications
Specifications
| Melting Point | 78°C |
| CAS | 12230-71-6 |
| Quantity | 1 kg |
| Molecular Formula | Ba(OH)2 · 8 H2O |
| MDL Number | MFCD00149152 |
| Synonym | Barium hydroxide octahydrate, Caustic baryta, Barium oxide hydrate octahydrate, Barium hydroxide octahydrate |
| Molecular Weight (g/mol) | 315.46 |
| Grade | Research |
| Density | 2.18 g/cm3 at 20°C |
| Chemical Name or Material | Barium Hydroxide Octahydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?