Learn More
Barium Hydroxide, European Pharmacopoeia Grade, Fisher Chemical™
BIOPHARMA
Product Code. 12980364 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. Quantity unitSize
12980364 1000 mL 1000mL
1 options
Product Code. 12980364
Brand: Fisher Chemical RP 030 1000
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 1000 mL
compliance-icons
Product Identifier
  • Barium Hydroxide Solution. European Pharmacopoeia Ref: 1009401 (RP 030)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.