Learn More
Barium Chloride, Supelco™
Product Code. 15887000
Change view
Click to view available options
Quantity:
50 g
500 g
Unit Size:
500g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15887000 500 g 500g
15854110 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15887000 Supplier Supelco Supplier No. 1017160500
 Restricted Item
Estimated Shipment: 19-11-2026
99.995 Suprapur™
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10361-37-2
Grade Research
Molecular Weight (g/mol) 208.23
Specifications
Boiling Point 1560°C/1013 hPa
Melting Point 963°C
Quantity 500 g
MDL Number MFCD00003445
Synonym Barium chloride, Barium dichloride, Barium chloride
Density 3.9 g/cm3 at 20°C
Chemical Name or Material Barium Chloride