Learn More
Barium Chloride Solution, 10%, Chem Lab™
Product Code. 11939162
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11939162
Brand: Chem-Lab CL02.0201.1000
Specifications
Quantity 1 L
compliance-icons
Product Identifier
  • Barium chloride 10% solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : BaCl2