Learn More
Thermo Scientific™ Barcoded Latch Racks for Matrix™ Storage Tubes
Product Code. 11620833
Change view
Click to view available options
For Use With (Equipment):
0.75mL tubes
1.0mL Screwtop Tubes
1.4mL tubes
500μL Screwtop Tubes
500μL Tubes
Quantity:
10 Case
5 Case
Unit Size:
Each
Pack of 10
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Quantity Unit Size
11620833 1.4mL tubes 10 Case Pack of 10
15543734 1.0mL Screwtop Tubes 5 Case Pack of 5
15553734 500μL Tubes 10 Case Pack of 10
15563734 500μL Screwtop Tubes 5 Case Pack of 5
15374297 0.75mL tubes 10 Case Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11620833 Supplier Thermo Scientific™ Supplier No. 4902
  • ONLINE EXCLUSIVE PRICE
    825.00 NOK
    670.36 NOK / Pack of 10
    Save 154.64 NOK
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Create custom arrays with empty ANSI-format racks featuring 2D and linear barcodes and human readable type on three sides and a 2D barcode on the base of the rack.
Enhance sample storage at low temperatures, including vapor phase Liquid Nitrogen, with Thermo Scientific™ Matrix™ barcoded latch racks. Barcoded Latch Racks feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack for the ultimate in sample tracking. Use in combination with Thermo Scientific 2D barcoded tubes, barcode readers, arrayers and capping equipment.
Superior Storage Rack Design
  • Storage in robot-friendly ANSI-footprint rack
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking regardless of your barcode system
  • Rack barcodes are compatible with our range of VisionMate readers
  • Hinged lid rests in “open” position on rack while conducting manual pipetting to prevent contamination of lid sitting on bench
Latch Racks Save Precious Space
  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Latch rack lid can be lifted robotically for tube access with high-throughput automated applications
TRUSTED_SUSTAINABILITY
Specifications
Description 1.4mL Latch Rack Lids
Capacity (Metric) 1.4 mL
For Use With (Equipment) 1.4mL tubes
Quantity 10 Case
Capacity (English) 0.0473 fl oz.
Product Type Barcoded Latch Rack