Description
- Manufactured from polypropylene
- Leak proof seal for integrity of storage
- Autoclavable vials and caps, can be re-sterilized
- Unique 1D, 2D bar-codes and human readable characters
- Features star shaped cavity in cap and is compatible with decapping equipment
- Locking base stabilizes tubes for automated handling
- Graduations and writing patch for manual use also
- Each tube is checked for clarity and readability of 2D code
Specifications
Specifications
| Closure Color | Orange |
| Color | Natural |
| Sterility | Sterile |
| Volume (Metric) | 2 mL |
| For Use With (Application) | Cell biology, cell banking, compound cryopreservation |
| No. per Case | 500 |
| No. per Pack | 50 |
| Thread Style | External |
| Base Style | Round, Self-standing |
| Barcode | 1D |
| Show More |