Learn More
JULABO™ Barbed Fittings
Product Code. 17108508
Change view
Click to view available options
For Use With (Equipment):
1 in. ID Male to G 1 in. Male
1.5 in. ID Male to G 1.5 in. Male
3/4 in. ID Male to G 3/4 in. Male
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17108508 3/4 in. ID Male to G 3/4 in. Male Each
17118508 1 in. ID Male to G 1 in. Male Each
17128508 1.5 in. ID Male to G 1.5 in. Male Each
3 options
Product Code. 17108508
Brand: JULABO™ 8890150
Barbed Fittings are fluid tubing adapters to convert male barbed fittings of various Internal Diameters (ID) to male G thread fittings with the same ID.
  • Fitting styles include:
    • 3/4 in. ID male to G 3/4 in. male
    • 1 in. ID male to G 1 in. male
    • 1 1/2 in. ID male to G 1 1/2 in. male
Specifications
For Use With (Equipment) 3/4 in. ID Male to G 3/4 in. Male
For Use With (Application) Fitting Adapter, Fluid Tubing
Quantity 2 Pk.