missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP31 Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | BAP31 |
|---|---|
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | CoraLite 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17741249
|
Proteintech
CL488-11200-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17629836
|
Proteintech
CL488-11200-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAP31 Polyclonal antibody apecifically detetects BAP31 antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.Specifications
| BAP31 | |
| Polyclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC014323 | |
| BCAP31 | |
| IgG | |
| Antigen affinity purification |
| Flow Cytometry | |
| CoraLite 488 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 10134 | |
| BAP31 fusion protein Ag1668 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title