missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BANP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561452
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PQALHYALAN VQQVQIHQIG EDGQVQ Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes a protein that binds to matrix attachment regions. The protein forms a complex with p53 and negatively regulates p53 transcription, and functions as a tumor suppressor and cell cycle regulator. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
BANP | |
Polyclonal | |
Unconjugated | |
BANP | |
AA408158; Banp; BEN domain-containing protein 1; BEND1; BTG3 associated nuclear protein; Btg3-associated nuclear protein; Protein BANP; scaffold/matrix-associated region-1-binding protein; Smar1; SMARBP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N9N5 | |
BANP | |
Recombinant protein corresponding to Human BANP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only