Learn More
Banoxantrone-d12 dihydrochloride, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Banoxantrone-d12 (AQ4N-d12) dihydrochloride is the deuterium labeled Banoxantrone dihydrochloride. Banoxantrone is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.
Specifications
Specifications
| Chemical Name or Material | Banoxantrone-d12 dihydrochloride |
| CAS | 1562066-98-1 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C22H18D12Cl2N4O6 |
| Technique | Cancer-programmed Cell Death |
| Color | Blue |
| Synonym | AQ4N-d12 dihydrochloride |
| SMILES | O=N(C([2H])([2H])[2H])(C([2H])([2H])[2H])CCNC1=CC=C(NCCN(C([2H])([2H])[2H])(C([2H])([2H])[2H])=O)C(C(C2=C(O)C=CC(O)=C23)=O)=C1C3=O.Cl.Cl |
| Molecular Weight (g/mol) | 529.48 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.