Learn More
Banoxantrone-d12 dihydrochloride, MedChemExpress
Product Code. 16401359
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 16401359
Brand: Medchem Express HY13562AS
MedChemExpress Banoxantrone-d12 (AQ4N-d12) dihydrochloride is the deuterium labeled Banoxantrone dihydrochloride. Banoxantrone is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.
Banoxantrone-d12 (AQ4N-d12) dihydrochloride is the deuterium labeled Banoxantrone dihydrochloride. Banoxantrone is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.
Specifications
Chemical Name or Material Banoxantrone-d12 dihydrochloride
CAS 1562066-98-1
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C22H18D12Cl2N4O6
Technique Cancer-programmed Cell Death
Color Blue
Synonym AQ4N-d12 dihydrochloride
SMILES O=N(C([2H])([2H])[2H])(C([2H])([2H])[2H])CCNC1=CC=C(NCCN(C([2H])([2H])[2H])(C([2H])([2H])[2H])=O)C(C(C2=C(O)C=CC(O)=C23)=O)=C1C3=O.Cl.Cl
Molecular Weight (g/mol) 529.48
Quantity 1 mg
Formula Weight 529.48
Percent Purity 98.02%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.