Learn More
BAN ORL 24, MedChemExpress
Product Code. 16495558
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16495558
Brand: Medchem Express HY13222
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress BAN ORL 24 is a potent and selective NOP receptor antagonist.
BAN ORL 24 is a potent and selective NOP receptor antagonist.
Specifications
Chemical Name or Material BAN ORL 24
CAS 1401463-54-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H37Cl2N3O2
Quantity 10 mg
Solubility Information DMSO : 200 mg/mL (394.86 mM; Need ultrasonic) ∣H2O : 100 mg/mL (197.43 mM; Need ultrasonic)
SMILES O=C([C@@H]1N(CC2=CC=CC=C2)CCC1)NCCCN3CCC4(CC3)OCC5=C4C=CC=C5.[H]Cl.[H]Cl
Molecular Weight (g/mol) 506.51
Formula Weight 506.51
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.