Learn More
Thermo Scientific Chemicals Ball pestle; clear plexiglas; 1/8in. diameter
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Item Description | Quantity: 25/Pk. |
| Diameter (English) Outer | 0.125 in. |
| Diameter (Metric) Outer | 3.17mm |
| Product Type | Ball Pestle |
| For Use With (Equipment) | Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT) |