Learn More
Sartorius Balance and Scale Weighing Pans
Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
Cubis 10mg and 100mg Balances
Unit Size:
Each
Description
Weighing pans for use with Sartorius Balances.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Bottom Shape | Flat |
| Color | Silver |
| Disposable | Reusable |
| Material | Stainless Steel |
| Shape | Rectangular |
| Sterility | Non-sterile |
| Type | Weighing Pan |
| For Use With (Application) | Weighing samples |
| For Use With (Equipment) | Cubis 10mg and 100mg Balances |