Learn More
Sartorius Balance Printers
Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
CPA Balance, Extended Balance, Moisture Analyzers
CPA Balance, Extended Balance, Moisture Analyzers. (For use w/ Cubis 1 MSE or MSA models.)
Dimensions (L x W x H):
215 x 155 x 78 mm
230 x 158 x 80 mm
Unit Size:
Each
Description
Transfer data automatically or manually. Sartorius™ Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
- Built-in real-time clock
- Built-in statistics program
- Counter for consecutive numbering of transactions can be activated
- High contrast LCD shows date or time as well as current transaction number
- Integrated rechargeable batteries with battery level indicator
- Ultrafast print head
- Runs on the balance's power; no separate power supply necessary
- Easy to change paper roll and ink ribbon cassette
Specifications
Specifications
| For Use With (Equipment) | CPA Balance, Extended Balance, Moisture Analyzers |
| Dimensions (L x W x H) | 215 x 155 x 78 mm |
| Length (Metric) | 215 mm |
| Width (Metric) | 155 cm |
| Height (Metric) | 78 mm |
| Includes | interface cable, Paper roll, four rechargeable b |
| Weight (Metric) | 0.9 kg |
| Type | Data Printer |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur