Learn More
Sartorius Balance Printers
GREENER_CHOICE
Product Code. 10239151 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
CPA Balance, Extended Balance, Moisture Analyzers
CPA Balance, Extended Balance, Moisture Analyzers. (For use w/ Cubis 1 MSE or MSA models.)
Dimensions (L x W x H):
215 x 155 x 78 mm
230 x 158 x 80 mm
Unit Size:
Each
Product Code. For Use With (Equipment) Dimensions (L x W x H) unitSize
10239151 CPA Balance, Extended Balance, Moisture Analyzers 215 x 155 x 78 mm Each
10506089 CPA Balance, Extended Balance, Moisture Analyzers. (For use w/ Cubis 1 MSE or MSA models.) 230 x 158 x 80 mm Each
2 options
Product Code. 10239151
Brand: Sartorius YDP200CE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Transfer data automatically or manually. Sartorius Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
Transfer data automatically or manually. Sartorius™ Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
  • Built-in real-time clock
  • Built-in statistics program
  • Counter for consecutive numbering of transactions can be activated
  • High contrast LCD shows date or time as well as current transaction number
  • Integrated rechargeable batteries with battery level indicator
  • Ultrafast print head
  • Runs on the balance's power; no separate power supply necessary
  • Easy to change paper roll and ink ribbon cassette
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CPA Balance, Extended Balance, Moisture Analyzers
Dimensions (L x W x H) 215 x 155 x 78 mm
Length (Metric) 215 mm
Width (Metric) 155 cm
Height (Metric) 78 mm
Includes interface cable, Paper roll, four rechargeable b
Weight (Metric) 0.9 kg
Type Data Printer