missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus Balance Compass CX621, 620g x 0.1g, External Calibration
Brand: Ohaus 30428209
597.21 NOK valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24121" to get your promotional price.
Balance Compass CX621, 620g x 0.1g, External Calibration
This item is not returnable.
View return policy
Description
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction