Learn More
J.T. Baker Baker-flex™, Silica Gel IB2-F, J.T. Baker™
Product Code. 15588294 Shop All J.T.Baker Products
Change view
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15588294 20 x 20 cm Each
15578294 20 x 20 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15588294 Supplier J.T. Baker Supplier No. 446704
  • ONLINE EXCLUSIVE PRICE
    9515.78 NOK
    8644.83 NOK / Pack of 1
    Save 870.95 NOK
    9% Off
 Restricted Item
Estimated Shipment: 09-04-2027
Flexible TLC Sheet
  • Material: High purity silica gel
  • Substrate: Lightweight, flexible, unbreakable polyester with abrasion-resistant coating
  • Layer thickness: 250 μm
  • Indicator: Fluorescent
  • Binder: Inert, does not char on sulfuric acid heat treatment
  • Features: Solvent-resistant, can be cut to size, can be written on, suitable for thin-layer chromatography
Specifications
Backing Material Polyester
Color White
Material Silica
Quantity 25 Sheets
Type TLC Sheet
Fluorescent Indicator F254
For Use With (Application) Thin-layer chromatography
Sorbent Silica Gel IB2-F (Fluorescent)
Thickness (Metric) 250 μm
Dimensions (L x W) 20 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm