Learn More
J.T. Baker Baker-flex™, Silica Gel IB2-F, J.T. Baker™
Flexible TLC Sheet
Brand:  J.T. Baker 4449-04
25165.50 NOK valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : CAS Number : 7631-86-9 Weight : 0.45000kg
 View more versions of this product
Product Code. 15578294
  • 26490.00 NOK / Each
Estimated Shipment: 29-05-2024
Description
Flexible TLC Sheet
Specifications
Specifications
White
25 Sheets
20 x 20 cm