Learn More
Interscience™ BagOpen 400 for Dilutors
Product Code. 16392226
Change view
Click to view available options
For Use With (Equipment):
DiluFlow, DiluFlow Pro, DiluFlow Elite
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16392226 DiluFlow, DiluFlow Pro, DiluFlow Elite Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16392226 Supplier Interscience™ Supplier No. 505001
  • ONLINE EXCLUSIVE PRICE
    7180.00 NOK
    7000.32 NOK / Pack of 1
    Save 179.68 NOK
    2% Off
 Restricted Item
Estimated Shipment: 19-01-2027
The BagOpen 400 opens bags for dilutors.
  • Stand for 400 mL bags
Specifications
Product Type Bag Stand
Autoclavable Not Autoclavable
Volume (Metric) 400 mL
For Use With (Application) Opening Bags
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite