missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26628-1-AP-150UL
This item is not returnable.
View return policy
Description
It has been hypothesizes that the BAG-5 protein will induce the death of nigral neurons through its predicted interaction with hsp70, which will cause increased protein aggregation and cell death by disinhibition of hsp70′s anti-apoptotic function. It is believed that BAG-5 will play an important role in the mechanisms of neuronal death. BAG-5 may also be of interest due to its possible role as a modulator of the hsp70/hsp40 chaperone axis or its possible interaction and coordination of localization/ modulation of other BAG containing proteins via BAG-BAG heterodimerization.Specifications
| BAG5 | |
| Polyclonal | |
| Unconjugated | |
| BAG5 | |
| BAG 5, BAG5, KIAA0873 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 366734, 70369, 9529 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5QJC9, Q8CI32, Q9UL15 | |
| Bag5 | |
| BAG5 Fusion Protein Ag24320 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction