Learn More
Fisherbrand™ Bacti-Loop Microsterilizer Loop Holder Attachment
Product Code. 31781852 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (English) Outside:
0.77 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Outside Unit Size
31781852 0.77 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781852
  • ONLINE EXCLUSIVE PRICE
    771.00 NOK
    692.27 NOK / Pack of 1
    Save 78.73 NOK
    10% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Helps to support and guide inoculation loops and tools in the Bacti-Loop Microsterilizer.
  • Supports inoculating loops and tools inside the chamber
  • Clamps on to Bacti-Loop body
Specifications
Diameter (English) Outside 0.77 in.
For Use With (Equipment) Bacti-Loop Microsterilizer
Product Type Loop Holder Attachment