Learn More
Cole-Parmer™ Bacti-Cinerator IV Infrared Sterilizer
Sterilize items by incinerating organic materials using infrared heat. Cole-Parmer™ Bacti-Cinerator IV Infrared Sterilizer requires only 5-7 seconds for optimum sterilization at 1500°F.
Brand: Cole-Parmer™ 01850-24
Additional Details : Weight : 0.80000kg
Description
Sterilize items by incinerating organic materials using infrared heat. Cole-Parmer™ Bacti-Cinerator IV Infrared Sterilizer requires only 5-7 seconds for optimum sterilization at 1500°F.
- Uses infrared heat to incinerate organic material deep within the ceramic funnel
- Sterilizes platinum loops and needles safely and conveniently
- Prevents infectious spatter and cross contamination
- Sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500°F
- Unit can be used in anaerobic chambers
- This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications
Specifications
Bacterial Incinerator/Infrared Sterilizer | |
50/60 Hz | |
Infrared heat |
1500°F | |
5 to 7 sec. | |
220 VAC |