Learn More
Cole-Parmer™ Coliform Test Kit
Product Code. 11737947 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11737947 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11737947 Supplier Cole-Parmer™ Supplier No. 9956100
  • ONLINE EXCLUSIVE PRICE
    617.00 NOK
    548.19 NOK / Pack of 1
    Save 68.81 NOK
    11% Off
View all promotions
Estimated Shipment: 12-02-2027
Test kit screens for coliform bacterial contamination
  • Simply pour sample into bag, shake to dissolve dechlorinating tablet, pour into the five glass tubes, and store tubes at room temperature
  • Color change and small bubble formation provide indication of bacterial presence
Specifications
Includes Sterile water sampling bag with dechlorinating tablet, five 10mL glass tubes, and five coliform test tablets.
Quantity 1 Each
Type Coliform Test Kit