Learn More
Testo™ 610 Compact Humidity/Temperature Meter with Backlit Display
Click to view available options
Temperature Range (Metric):
-10°C to +50°C
Unit Size:
Each
Description
- Easy temperature and humidity measurement
- Calculation of dew point and wet bulb temperature
- Illuminated display
- Capacitive humidity sensor with long-term stability
Specifications
Specifications
| Humidity Range | 0 to 100% |
| Resolution | Humidity: 0.1 %RH |
| Accuracy | NTC: ±0.5°C; Humidity: ±2.5 %RH (5 to 95 %RH) |
| Temperature Range (Metric) | -10°C to +50°C |
| Temperature Accuracy | ±0.5°C |
| Temperature Resolution | 0.1°C |
| Dimensions (L x W x H) | 119 x 46 x 25 mm (incl. protective cap) |
| Weight (Metric) | 90 g |