missing translation for 'onlineSavingsMsg'
Learn More

B-myc Polyclonal Antibody, Invitrogen™

Product Code. 15896323
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
15896323 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 15896323

Brand: Invitrogen PA567911

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Seems to act as an inhibitor of cellular proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B-myc
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Bmyc
Gene Accession No. Q6P8Z1, P15063
Gene Alias Protein B-Myc, Mycb, Bmyc
Gene Symbols Bmyc
Host Species Rabbit
Immunogen Synthesized peptide derived from human Mycb.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107771, 311807
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.