Learn More
Vitlab™ Büchner Funnels
Filter out impurities with Buchner funnel. Vitlab™ Büchner funnels are exceptional for filtration and removing impurities. The upper and lower part of the funnel is detachable to facilitate cleaning.
Brand:  Vitlab™ 80437
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10778101
  • 174.00 NOK / Each
Estimated Shipment: 12-06-2024
Description
  • Polypropylene construction
  • >
  • Upper and lower parts are detachable
Specifications
Specifications
40 mL
42.5mm
95mm
Polypropylene
1.2mm