Learn More
Brady™ B-7606 Paper Thermal Transfer Labels
Product Code. p-7225740
Click to view available options
Height (Metric):
101.6 mm
127 mm
152.4 m
19.1 mm
38.1 mm
50.8 mm
76.2 mm
Shape:
Rectangle
Square
Vertical Repeat (Metric):
22.2 mm
41.3 mm
54 mm
79.4 mm
104.8 mm
130.2 mm
155.6 mm
Width (Metric):
57.2 mm
101.6 mm
Unit Size:
Each
Product Code. 11523125
Brand: Brady™ 236733
Only null left
Add to basket
Add to basket
Printable, paper labels for Thermal Transfer Printers.
  • Permanent acrylic adhesive
  • 25 mm small core
  • Glossy finish
Specifications
Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Color White
Finish Gloss
Material Paper (B-7606)
Print Technology Thermal Transfer
Sterility Non-sterile
For Use With (Application) Warehouse Marking
For Use With (Equipment) BBP™11, BBP™12, and THT 76 mm Core Printers
Labels per Row 1
Quantity 21,360 Labels
Shape Rectangle
Temperature Range (Metric) -40°C to +50°C
Thickness (Metric) 0.081 mm
Vertical Repeat (Metric) 22.2 mm
Width (Metric) 57.2 mm
Height (Metric) 19.1 mm
Alternate Part Number BPT-722-075