Learn More
Brady™ B-437 Polyvinylfluoride Self-extinguishing Thermal Transfer Labels
Product Code. 11553744
Click to view available options
Color:
White
Yellow
Height (Metric):
6.4 mm
9.5 mm
12.7 mm
Labels per Row:
2
3
4
Width (Metric):
15.2 mm
19.1 mm
22.9 mm
25.4 mm
31.8 mm
Unit Size:
1 roll
Product Code. 11553744
Brand: Brady™ 030163
Only null left
Add to basket
Add to basket
Printable, self-extinguishing labels for Thermal Transfer Printers.
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
  • Self-extinguishing
Specifications
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Die-cut
Material PVF
Product Type Label
Sterility Non-sterile
For Use With (Application) Circuit Board Labeling
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Horizontal Repeat (Metric) 20.3 mm
Labels per Row 4
Quantity 10,000 Labels
Shape Rectangle
Temperature Range (English) -94°F to 275°F
Temperature Range (Metric) -70°C to +135°C
Thickness (English) 0.0036 in.
Thickness (Metric) 0.091 mm
Vertical Repeat (Metric) 8.9 mm
Dimensions (LxW) 19.05 mm (W) x 6.35 mm (H)
Length (English) 0.25 in.
Width (English) 0.75 in.
Width (Metric) 19.1 mm
Width (Metric) Printable Area 19.1 mm
Height (English) 0.25 in.
Height (Metric) 6.4 mm
Alternate Part Number THT-1-437W-10