Learn More
Brady™ B-423 Polyester Thermal Transfer Labels
Product Code. 11589053
Change view
Click to view available options
Height (Metric):
3.2 mm
4.8 mm
5.1 mm
6.4 mm
9.5 mm
9.7 mm
10.2 mm
11.1 mm
12.7 mm
19.1 mm
31.8 mm
Labels per Row:
1
2
3
4
5
7
12
Vertical Repeat (Metric):
6.4 mm
7.3 mm
7.6 mm
8.9 mm
9.5 mm
12.1 mm
12.7 mm
13.6 mm
15.2 mm
22.2 mm
34.9 mm
Width (Metric):
6.4 mm
9.5 mm
12.7 mm
16.5 mm
17.8 mm
19.1 mm
22.9 mm
25.4 mm
27.9 mm
31.8 mm
34.9 mm
38.1 mm
50.8 mm
69.9 mm
Unit Size:
1 roll
Each
Product Code. Height (Metric) Labels per Row Vertical Repeat (Metric) Width (Metric) unitSize
11589053 5.1 mm 1 7.6 mm 12.7 mm 1 roll
11593764 3.2 mm 2 6.4 mm 38.1 mm 1 roll
11558254 4.8 mm 1 7.3 mm 25.4 mm 1 roll
11508264 5.1 mm 1 7.6 mm 16.5 mm 1 roll
11583704 5.1 mm 4 7.6 mm 16.5 mm 1 roll
11598254 6.4 mm 1 8.9 mm 19.1 mm 1 roll
11528264 6.4 mm 1 8.9 mm 22.9 mm 1 roll
11599053 6.4 mm 1 8.9 mm 25.4 mm 1 roll
11568254 6.4 mm 1 8.9 mm 31.8 mm 1 roll
11578254 6.4 mm 1 8.9 mm 34.9 mm 1 roll
11518264 6.4 mm 1 8.9 mm 50.8 mm 1 roll
11543714 6.4 mm 2 9.5 mm 31.8 mm 1 roll
11563784 6.4 mm 2 9.5 mm 34.9 mm 1 roll
11573704 6.4 mm 3 8.9 mm 22.9 mm 1 roll
11513714 6.4 mm 12 9.5 mm 6.4 mm 1 roll
11523804 9.5 mm 2 12.1 mm 31.8 mm 1 roll
15751427 9.5 mm 3 12.1 mm 25.4 mm 1 roll
11543814 9.5 mm 4 12.1 mm 17.8 mm 1 roll
11523784 9.5 mm 7 12.1 mm 9.5 mm 1 roll
11573794 9.7 mm 2 12.7 mm 27.9 mm 1 roll
16677865 10.2 mm 1 12.7 mm 50.8 mm Each
11523714 11.1 mm 5 13.6 mm 12.7 mm 1 roll
11543804 12.7 mm 1 15.2 mm 50.8 mm 1 roll
11553704 19.1 mm 2 22.2 mm 38.1 mm 1 roll
11583794 19.1 mm 4 22.2 mm 19.1 mm 1 roll
16667865 31.8 mm 1 34.9 mm 69.9 mm Each
26 options
Product Code. 11589053
Brand: Brady™ 018654
Only null left
Add to basket
Add to basket
Printable, polyester labels for Thermal Transfer Printers.
  • Permanent adhesive
  • For smooth surfaces
Specifications
Autoclavable Autoclavable
Certifications/Compliance UL, CSA and AGA Approved
Adhesive Type Permanent Acrylic
Color White
Finish Gloss
Format Die-cut
Material Polyester (B-423)
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory Labeling
For Use With (Equipment) BBP11;BBP12;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
Labels per Row 1
Min. Operating Temperature (Metric) -70°C
Model THT-97-423-10
No. per Pack 10000 Label(s) / Roll
Quantity 10,000 Labels
Shape Rectangle
Size 12.70 mm (W) x 5.08 mm (H)
Temperature Range (English) -94°F to +230°F
Temperature Range (Metric) -70°C to +110°C
Thickness (English) 0.003 in
Thickness (Metric) 0.076 mm
Vertical Repeat (Metric) 7.6 mm
Dimensions (LxW) 12.70 mm (W) x 5.08 mm (H)
Width (English) 0.5 in
Width (Metric) 12.7 mm
Width (English) Printable Area 12.70 mm
Height (English) 0.2 in
Height (Metric) 5.1 mm
Alternate Part Number THT-97-423-10