Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ B-423 Polyester Thermal Transfer Labels
Brady™ B-423 Polyester Thermal Transfer Labels
Product Code. 11543804
Change view
Click to view available options
Height (Metric):
3.2 mm
4.8 mm
5.1 mm
6.4 mm
9.5 mm
9.7 mm
10.2 mm
11.1 mm
12.7 mm
19.1 mm
31.8 mm
Labels per Row:
1
2
3
4
5
7
12
Vertical Repeat (Metric):
6.4 mm
7.3 mm
7.6 mm
8.9 mm
9.5 mm
12.1 mm
12.7 mm
13.6 mm
15.2 mm
22.2 mm
34.9 mm
Width (Metric):
6.4 mm
9.5 mm
12.7 mm
16.5 mm
17.8 mm
19.1 mm
22.9 mm
25.4 mm
27.9 mm
31.8 mm
34.9 mm
38.1 mm
50.8 mm
69.9 mm
Unit Size:
1 roll
Each
| Product Code. | Height (Metric) | Labels per Row | Vertical Repeat (Metric) | Width (Metric) | unitSize |
|---|---|---|---|---|---|
| 11543804 | 12.7 mm | 1 | 15.2 mm | 50.8 mm | 1 roll |
| 11593764 | 3.2 mm | 2 | 6.4 mm | 38.1 mm | 1 roll |
| 11558254 | 4.8 mm | 1 | 7.3 mm | 25.4 mm | 1 roll |
| 11589053 | 5.1 mm | 1 | 7.6 mm | 12.7 mm | 1 roll |
| 11508264 | 5.1 mm | 1 | 7.6 mm | 16.5 mm | 1 roll |
| 11583704 | 5.1 mm | 4 | 7.6 mm | 16.5 mm | 1 roll |
| 11598254 | 6.4 mm | 1 | 8.9 mm | 19.1 mm | 1 roll |
| 11528264 | 6.4 mm | 1 | 8.9 mm | 22.9 mm | 1 roll |
| 11599053 | 6.4 mm | 1 | 8.9 mm | 25.4 mm | 1 roll |
| 11568254 | 6.4 mm | 1 | 8.9 mm | 31.8 mm | 1 roll |
| 11578254 | 6.4 mm | 1 | 8.9 mm | 34.9 mm | 1 roll |
| 11518264 | 6.4 mm | 1 | 8.9 mm | 50.8 mm | 1 roll |
| 11543714 | 6.4 mm | 2 | 9.5 mm | 31.8 mm | 1 roll |
| 11563784 | 6.4 mm | 2 | 9.5 mm | 34.9 mm | 1 roll |
| 11573704 | 6.4 mm | 3 | 8.9 mm | 22.9 mm | 1 roll |
| 11513714 | 6.4 mm | 12 | 9.5 mm | 6.4 mm | 1 roll |
| 11523804 | 9.5 mm | 2 | 12.1 mm | 31.8 mm | 1 roll |
| 15751427 | 9.5 mm | 3 | 12.1 mm | 25.4 mm | 1 roll |
| 11543814 | 9.5 mm | 4 | 12.1 mm | 17.8 mm | 1 roll |
| 11523784 | 9.5 mm | 7 | 12.1 mm | 9.5 mm | 1 roll |
| 11573794 | 9.7 mm | 2 | 12.7 mm | 27.9 mm | 1 roll |
| 16677865 | 10.2 mm | 1 | 12.7 mm | 50.8 mm | Each |
| 11523714 | 11.1 mm | 5 | 13.6 mm | 12.7 mm | 1 roll |
| 11553704 | 19.1 mm | 2 | 22.2 mm | 38.1 mm | 1 roll |
| 11583794 | 19.1 mm | 4 | 22.2 mm | 19.1 mm | 1 roll |
| 16667865 | 31.8 mm | 1 | 34.9 mm | 69.9 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Permanent adhesive
- For smooth surfaces
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | UL, CSA and AGA Approved |
| Adhesive Type | Permanent Acrylic |
| Color | White |
| Finish | Gloss |
| Format | Die-cut |
| Material | Polyester (B-423) |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory Labeling |
| For Use With (Equipment) | BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500 |
| Labels per Row | 1 |
| Min. Operating Temperature (Metric) | -70°C |
| Model | THT-53-423-3 |
| No. per Pack | 3000 Label(s) / Roll |
| Quantity | 3000 Labels |
| Shape | Rectangle |
| Size | 50.80 mm (W) x 12.70 mm (H) |
| Temperature Range (English) | -94°F to +230°F |
| Temperature Range (Metric) | -70°C to +110°C |
| Thickness (English) | 0.003 in |
| Thickness (Metric) | 0.076 mm |
| Vertical Repeat (Metric) | 15.2 mm |
| Dimensions (LxW) | 50.80 mm (W) x 12.70 mm (H) |
| Width (English) | 2 in |
| Width (Metric) | 50.8 mm |
| Width (English) Printable Area | 50.80 mm |
| Height (English) | 0.5 in |
| Height (Metric) | 12.7 mm |
| Alternate Part Number | THT-53-423-3 |
| Show More Show Less |