Learn More
Azomethine H, MP Biomedicals™
Product Code. 11417532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Product Code. Quantity unitSize
11417532 1 g 1g
11427532 5 g 5g
11437532 10 g 10g
3 options
Product Code. 11417532
Brand: MP Biomedicals 0215416901
Chemical Identifiers
CAS 5941-07-1
Molecular Formula C17H14NNaO9S2
Molecular Weight (g/mol) 463.41
MDL Number MFCD00149588
InChI Key BEDIAJJRAYEWBI-TTWKNDKESA-M
Synonym 8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate
ChEBI CHEBI:368
Specifications
Assay Percent Range 1.04
Extinction Coefficient Extinction Coefficient max: 237nm C=0.01% ≥50, 000 (Lit.)
SMILES O.[Na+].OC1=C2C(N\C=C3/C=CC=CC3=O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S([O-])(=O)=O
IUPAC Name sodium 5-hydroxy-4-({[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)-7-sulfonaphthalene-2-sulfonate hydrate
PubChem CID 443157
Formula Weight 423.4
Color Orange
Quantity 1 g
Physical Form Powder
Chemical Name or Material Azomethine H, Chemistry