Learn More
Azlon™ HDPE weighing scoop
Light weight and chemical resistant
Brand: Azlon FWC324
Additional Details : Weight : 7.30000kg
Description
- Lightweight HDPE weighing boats
- Chemical resistant
Specifications
35 mL | |
Boat | |
100 mm |
HDPE | |
Weighing Scoop |