Learn More
Azlon™ USA OSHA Vented Wide Mouth Wash Bottles
Description
- Indelibly printed in 3 languages (English, French and Spanish)
- Choose from five solvent labels all including the GHS, NFPA diamond, PPE pictograms, risk and safety information
- Fitted with color coded one-piece PP closures that maintain their shape in use
- Closures feature precision jet-tip aperture for fine control
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Plastic |
| Quantity | 5/Pk. |
| Chemical Label | Ethanol |
| Closure Type | Vapor-venting |