Learn More
Azlon™ Thistle Shaped Funnel with Detachable Long Stem
Product Code. 10365250
Change view
Click to view available options
Length (Metric) Stem:
25 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Stem Unit Size
10365250 25 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10365250 Supplier Azlon Supplier No. FWC220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made of polypropylene
  • Thistle shaped funnel with detachable long stem
  • Stem dimensions: 6mm O. D. x 4mm I. D. x 25mm long
  • Stem can be cut to size
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 34mm
Diameter (Metric) Stem 6mm
Material Polypropylene
Length (Metric) Stem 25 mm
Product Type Thistle funnel