Learn More
Azlon™ Polystyrene Specimen Container with Polyethylene Lid
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
100 mL
150 mL
200 mL
250 mL
Diameter (Metric):
32 mm
36 mm
51 mm
65 mm
Unit Size:
Pack of 10
Pack of 5
Description
- Clear polystyrene containers with maximum sample visibility
- Polyethylene airtight snap-on lids for quick easy use
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Material | Polystyrene |
| Sterility | Non-sterile |
| Capacity (Metric) | 100 mL |
| Closure Type | Snap Cap |
| Color | Clear |
| Diameter (Metric) | 51 mm |
| Height (Metric) | 60 mm |
| Includes | Polyethylene lid |
| Show More |