Learn More
Azlon™ Polypropylene Test Tube Rack
Product Code. p-8001848
Change view
Click to view available options
Color:
Blue
White
Positions:
21
32
40
55
84
Diameter (Metric) Opening:
13 mm
18 mm
21 mm
26 mm
31 mm
Dimensions (L x W x H):
265 x 126 x 75 mm
265 x 126 x 88 mm
Unit Size:
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Positions Diameter (Metric) Opening Dimensions (L x W x H) unitSize
11387794 Blue 40 21 mm 265 x 126 x 75 mm Pack of 5
12531406 Blue 32 26 mm 265 x 126 x 88 mm Pack of 5
11357794 Blue 84 13 mm 265 x 126 x 75 mm Pack of 5
11367794 Blue 55 18 mm 265 x 126 x 75 mm Pack of 5
11933849 Blue 21 31 mm 265 x 126 x 88 mm Pack of 5
11377794 White 40 21 mm 265 x 126 x 75 mm Pack of 5
11397794 White 32 26 mm 265 x 126 x 88 mm Pack of 5
11347794 White 84 13 mm 265 x 126 x 75 mm Pack of 5
11971528 White 55 18 mm 265 x 126 x 75 mm Pack of 5
11307804 White 21 31 mm 265 x 126 x 88 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11387794 Supplier Azlon Supplier No. RPB021P
Only null left
Add to basket
Add to basket
Economical and sturdy test tube racks supplied in self assembly flat packs
Economical and sturdy test tube racks supplied in self assembly flat packs
  • Autoclavable at 121°C
  • Sinks in water baths
  • Alpha numeric place coding
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Blue
Holds Tubes (20 to 21 mm)
Material Polypropylene
Positions 40
Dimensions (L x W x H) 265 x 126 x 75 mm
Diameter (Metric) Opening 21 mm
Product Type Tube Rack
Diameter (Metric) Inner 21 mm