Learn More
Azlon™ Polypropylene Powder Funnels
Product Code. 12054638
Change view
Click to view available options
Diameter (Metric)Top:
65mm
80mm
100mm
120mm
150mm
290mm
420mm
Diameter (Metric) Stem:
18mm
22mm
26mm
30mm
35mm
36mm
48mm
Height (Metric):
69 mm
74 mm
90 mm
106 mm
135 mm
208 mm
270 mm
Unit Size:
Each
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Height (Metric) unitSize
12054638 100mm 26mm 90 mm Pack of 5
12571506 120mm 30mm 106 mm Pack of 5
10589730 150mm 36mm 135 mm Pack of 5
12030783 290mm 35mm 208 mm Each
12040783 420mm 48mm 270 mm Each
12074628 65mm 18mm 69 mm Pack of 5
12561506 80mm 22mm 74 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12054638 Supplier Azlon Supplier No. FWC206
Only null left
Add to basket
Add to basket
Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.
  • Lightweight polypropylene powder funnel
  • Unbreakable in normal use
  • Parallel stem minimises 'bridging' of powder to improve flow
  • The two largest sizes have vertical-sided tops
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 100mm
Diameter (Metric) Stem 26mm
Material Polypropylene
Height (Metric) 90 mm