Learn More
Azlon™ Polyethylene Stoppers
Product Code. 12987326
Change view
Click to view available options
Standard Taper Joint:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Unit Size:
Pack of 10
Product Code. Standard Taper Joint unitSize
12987326 12/21 Pack of 10
12977326 10/19 Pack of 10
11317804 14/23 Pack of 10
11327804 19/26 Pack of 10
12997326 29/32 Pack of 10
12907336 34/35 Pack of 10
11337804 24/29 Pack of 10
7 options
Product Code. 12987326
Brand: Azlon™ TRF592
Only null left
Add to basket
Add to basket
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. Azlon™ Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. AzlonTM Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
  • Manufactured from polyethylene
  • Available in white and red
  • Suitable for use with both Azlon and glass volumetric ware
  • Enlarged head provides a good grip and protects glassware
  • Ribbed cone ensures a secure leak free seal
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Glass or Plastic volumetric flasks and measuring cylinders etc
Standard Taper Joint 12/21
Color White, Red
Material Polyethylene
Product Type Tapered Stopper
Shape Ribbed Cone